Learn More
Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Tubing
Product Code. 11364645
Click to view available options
Diameter (English) Inner:
3/32 in.
5/16 in.
Diameter (English) Outer:
5/32 in.
7/16 in.
Overall Length (English):
50 ft.
Unit Size:
15m
Product Code. 11364645
Brand: Thermo Scientific™ 80001090
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Perform a range of non-medical applications with this 180 clear plastic PVC tubing, which resists hardening and discoloration.
Perform a range of non-medical applications with Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Tubing, which resists hardening and discoloration from a wide range of chemicals. Manufactured from the highest-grade resins and special plasticizers, this tubing is available in 50, 100, 250 and 500' cases. Meets Food Additives Amendment of the Federal Food, Drug, and Cosmetic Act; also USDA and U.S. Pharmacopeia Class VI requirements for plastic materials.
Features:
  • Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from the highest-grade resins and plasticizers
  • Resists hardening and discoloration from a wide range of chemicals–demonstrates dimensional stability
  • Equivalent to Tygon™ R-3603 tubing
  • Marked with I.D. and regulatory compliance as well as at 12 in. intervals for easy identification and measurement (except as noted below)
  • Durometer hardness is Shore A, 55
  • Autoclavable/transparent
Note: The following sizes are not printed: 8000-0002, -0004, -0006, -0010, -0025 and -1090
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Material is Food-Grade and complies with USP VI and USDA regulations.
Diameter (English) Inner 5/16 in.
Material PVC
Diameter (English) Outer 7/16 in.
Overall Length (English) 50 ft.
Thickness (English) 1/16 in.
Packaging Quantity 1 piece of tubing 10 ft in length
Maximum Pressure 30 psig at 73°F (23°C), 15psig at 125°F (52°C)
Not intended for medical use. Nalgene 180 and 980 PVC tubing contain DEHP [Bis (2-ethylhexyl)] phthalate, a commonly used plasticizer. It is known to the state of California Environmental Agency that this chemical causes cancer or reproductive toxicity.