Learn More
Thermo Scientific™ Nalgene™ HDPE Quick Disconnects
Product Code. 10345751
Click to view available options
For Use With:
1/4 to 5/16 in. I.D.
3/8 to 7/16 in. I.D.
Unit Size:
Pack of 12
Pack of 72
Product Code. 10345751
Brand: Thermo Scientific™ 61500010
Only null left
Add to basket
Add to basket
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ Quick Disconnects.
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ HDPE Quick Disconnects, which allow tubulations to slide together snugly with a twist.
Two tapered tubulations fit tightly together with male/female connection, allowing for rapid joining or separation of apparatus lines.
TRUSTED_SUSTAINABILITY
Specifications
Material HDPE
For Use With 1/4 to 5/16 in. I.D.
Packaging Quantity 12/Pk., 72/Cs.
Product Type Quick Disconnect
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.