Learn More
Thermo Scientific™ Nalgene™ HDPE Quick Disconnects
Product Code. 10575591
Click to view available options
For Use With:
1/4 to 5/16 in. I.D.
3/8 to 7/16 in. I.D.
Unit Size:
Pack of 12
Pack of 72
Product Code. 10575591
Brand: Thermo Scientific™ 61500020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ Quick Disconnects.
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ HDPE Quick Disconnects, which allow tubulations to slide together snugly with a twist.
Two tapered tubulations fit tightly together with male/female connection, allowing for rapid joining or separation of apparatus lines.
TRUSTED_SUSTAINABILITY
Specifications
Material HDPE
For Use With 3/8 to 7/16 in. I.D.
Packaging Quantity 12/Pk., 72/Cs.
Product Type Quick Disconnect
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.