Learn More
Naloxone Hydrochloride Dihydrate, Mikromol™
Description
Naloxone Hydrochloride Dihydrate
Specifications
Specifications
| Chemical Name or Material | Naloxone hydrochloride dihydrate |
| CAS | 51481-60-8 |
| Analyte or Component Names | Naloxone hydrochloride dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H21 N O4 . Cl H . 2 H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Naloxone Hydrochloride Dihydrate, Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propenyl)-, hydrochloride, dihydrate, (5alpha)- |
| InChI Formula | InChI=1S/C19H21NO4.ClH.2H2O/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11;;;/h2-4,14,17,21,23H,1,5-10H2;1H;2*1H2/t14-,17+,18+,19-;;;/m1.../s1 |
| SMILES | O.O.Cl.Oc1ccc2C[C@H]3N(CC=C)CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O |
| IUPAC Name | (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;dihydrate;hydrochloride |
| Show More |