missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NALP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584063
This item is not returnable.
View return policy
Description
Immunogen sequence: LNEDELKSFK SLLWAFPLED VLQKTPWSEV EEADGKKLAE ILVNTSSENW IRNATVNILE EMNLTELCKM AK Highest antigen sequence indentity to the following orthologs: Rat - 33%, Mouse - 35%.
NOD proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins. NALP7, also known as PYPAF3, a member of the related PYRIN-containing APAF1-like proteins (PYPAFs/NALPs) is thought to play a crucial role in cell proliferation. NALP7 has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. It is expressed in numerous tissues including uterus and ovary, with low levels in heart and brain. NALP7 inhibits caspase-1-dependent interleukin-1-b secretion and is a feedback regulator of interleukin-1-b secretion. Defects in the NALP7 gene are known to cause the formation of a hydatidiform mole (HYDM) and reduce the growth of carcinoma cell lines.
Specifications
| NALP7 | |
| Polyclonal | |
| Unconjugated | |
| NLRP7 | |
| CLR19.4; HYDM; NACHT, leucine rich repeat and PYD containing 7; NACHT, LRR and PYD containing protein 7; NACHT, LRR and PYD domains-containing protein 7; NALP7; NLR family pyrin domain containing 7; NLR family, pyrin domain containing 7; NLRP7; NOD12; Nucleotide-binding oligomerization domain protein 12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 7; PAN7; PYPAF3; PYRIN-containing APAF1-like protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 199713 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WX94 | |
| NLRP7 | |
| Recombinant protein corresponding to Human NLRP7. Recombinant protein control fragment (Product #RP-99144). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction