missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-L-ornithine, 98%, Thermo Scientific Chemicals

Catalog Number p-7054670
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2640-58-6
C13H18N2O4
266.297
MFCD00038200
ZYGRWJVRLNJIMR-NSHDSACASA-N
z-orn-oh, cbz-l-ornithine, cbz-orn-oh, l-ornithine, n2-phenylmethoxy carbonyl, 2s-5-amino-2-benzyloxy carbonyl amino pentanoic acid, s-5-amino-2-benzyloxycarbonylamino pentanoic acid, s-5-amino-2-benzyloxy carbonyl amino pentanoic acid, pubchem18987, carbobenzyloxy-l-ornithine, na-carbobenzoxy-l-ornithine
7010490
(2S)-5-amino-2-(phenylmethoxycarbonylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCCN)C(=O)O
This item is not returnable. View return policy
2640-58-6
C13H18N2O4
266.297
MFCD00038200
ZYGRWJVRLNJIMR-NSHDSACASA-N
z-orn-oh, cbz-l-ornithine, cbz-orn-oh, l-ornithine, n2-phenylmethoxy carbonyl, 2s-5-amino-2-benzyloxy carbonyl amino pentanoic acid, s-5-amino-2-benzyloxycarbonylamino pentanoic acid, s-5-amino-2-benzyloxy carbonyl amino pentanoic acid, pubchem18987, carbobenzyloxy-l-ornithine, na-carbobenzoxy-l-ornithine
7010490
(2S)-5-amino-2-(phenylmethoxycarbonylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCCN)C(=O)O

Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.

Solubility
Soluble in 0.1N NaOG (1 mg/ml-clear, colorless solution). Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Solubility Information Soluble in 0.1N NaOG (1mg/ml-clear,colorless solution). Slightly soluble in water.
Formula Weight 266.29
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material Nalpha-Benzyloxycarbonyl-L-ornithine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'