missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-L-arginine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7055112
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
35897-34-8
C11H23ClN4O4
310.78
MFCD00065552
HDELGKMVZYHPPB-RZUXQYDTNA-N
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, boc-l-arg-ohhclh2o
2729004
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O
This item is not returnable. View return policy
35897-34-8
C11H23ClN4O4
310.78
MFCD00065552
HDELGKMVZYHPPB-RZUXQYDTNA-N
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, boc-l-arg-ohhclh2o
2729004
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Beilstein 3658939
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride
Formula Weight 310.78
Percent Purity 98%
Quantity 5 g
Physical Form Powder
Chemical Name or Material Nalpha-Boc-L-arginine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'