missing translation for 'onlineSavingsMsg'
Learn More

Nalpha,Nepsilon-Di-Boc-L-lysine 4-nitrophenyl ester, 98%, Thermo Scientific Chemicals

Catalog Number p-7058172
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2592-19-0
C22H33N3O8
467.52
MFCD00076961
LYUXBTAUKJETMS-AGDOHHJYNA-N
boc-lys boc-onp, ambotzbaa5780, 4-nitrophenyl 2s-2,6-bis tert-butoxycarbonyl amino hexanoate, n2,n6-di-boc-l-lysine 4-nitrophenyl ester, n-alpha-n-epsilon-di-t-butyloxycarbonyl-l-lysine p-nitrophenyl ester, l-lysine, n2,n6-bis 1,1-dimethylethoxy carbonyl-, 4-nitrophenyl ester
13455305
(4-nitrophenyl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O
This item is not returnable. View return policy
2592-19-0
C22H33N3O8
467.52
MFCD00076961
LYUXBTAUKJETMS-AGDOHHJYNA-N
boc-lys boc-onp, ambotzbaa5780, 4-nitrophenyl 2s-2,6-bis tert-butoxycarbonyl amino hexanoate, n2,n6-di-boc-l-lysine 4-nitrophenyl ester, n-alpha-n-epsilon-di-t-butyloxycarbonyl-l-lysine p-nitrophenyl ester, l-lysine, n2,n6-bis 1,1-dimethylethoxy carbonyl-, 4-nitrophenyl ester
13455305
(4-nitrophenyl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 467.51
Percent Purity 98%
Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine 4-nitrophenyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'