Learn More
Invitrogen™ NAMPT Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA11045
Product Code. 11535892
  • € 570.00 / 100µg
Description
PA1-1045 detects Visfatin (PBEF) in human samples. PA1-1045 has been successfully used in Western blot procedures. In Western blot analysis of human adipose tissue lysate this antibody detects a ~52 kDa protein representing Visfatin. The PA1-1045 immunogen is a synthetic peptide corresponding to residues S (472) F D E I R K N A Q L N I E L E A A H H (491) of human Visfatin.
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NAMPT
Polyclonal
Unconjugated
NAMPT
1110035O14Rik; AI314458; AI480535; DKFZp666B131; EC 2.4.2.12; hypothetical protein; MGC117256; NAmPRTase; NAMPT; nampt1; nampta; NAMPTP1; Nicotinamide phosphoribosyltransferase; nicotinamide phosphoribosyltransferase 1; nicotinamide phosphoribosyltransferase a; nicotinamide phosphoribosyltransferase pseudogene 1; PBEF; Pbef1; pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; pre-B-cell colony-enhancing factor 1; Pre-B-cell colony-enhancing factor 1 homolog; pre-B-cell colony-enhancing factor 1 precursor; VF; VISFATIN
Rabbit
Antigen affinity chromatography
RUO
10135
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1 mg/mL BSA and 0.05% sodium azide
P43490
NAMPT
Synthetic peptide corresponding to residues S (472) F D E I R K N A Q L N I E L E A A H H (491) of human Visfatin.