Learn More
Melford NANA
Description
Substrate for N-Acetylneuraminic acid aldolase.
Substance Name: n-Acetylneuraminic acid, Synthetic Sialic Acid
CAS Number: 131-48-6
Molecular Formula: C11H19NO9
Molecular Weight: 309.2
Chemical Purity:>99%
Soluble in:Water
Store at:-20°CSpecifications
Specifications
| CAS | 131-48-6 |
| Molecular Formula | C11H19NO9 |
| Quantity | 25 mg |
| Molecular Weight (g/mol) | 309.3 |