Learn More
Thermo Scientific™ nanoCell Accessory
Product Code. 11741141 Shop All Thermo Scientific Products
Click to view available options
Type:
NanoCell, 1.0mm pathlength
Unit Size:
Each
Product Code. 11741141
Brand: Thermo Scientific™ 222227800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ nanoCell accessory provides accurate and sensitive measurements of small volume samples.
Thermo Scientific™ nanoCell accessory provides accurate and sensitive measurements of small volume samples. The nanoCell accessory allows you to measure samples volumes from less than a microliter to milliliters using the same spectrophotometer. Use large volume or disposable cuvettes for some assays and the nanoCell accessory for microliter samples.With the nanoCell accessory, you have all the features of a quality Thermo Scientific spectrophotometer plus the benefit of small volume measurements.
  • No alignment required; just insert the nanoCell, load your sample and get accurate results
  • Removable accessory makes the switch between large and small volume measurements fast and easy
  • Use between 0.7 and 5.0μL of sample
  • Can measure DNA concentration measurements from 4.0 to 4500μg/mL
  • Quick cleaning with a laboratory tissue allows high sample throughput and excellent sample carry-over performance
Recommended for:
Life science
TRUSTED_SUSTAINABILITY
Specifications
Type NanoCell, 1.0mm pathlength
For Use With (Equipment) BioMate 3, GENESYS 10 Bio
Pathlength 1.0 mm