Learn More
Macherey Nagel NANOCOLOR PTFE pressure digestion vessel 20ml
Brand:  Macherey Nagel 91643
675.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
NANOCOLOR PTFE pressure digestion vessel 20ml
Product Code. 10013141
  • € 794.00 / Each
Only null left
Estimated Shipment: 03-09-2025