missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanog Antibody (1E6C4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47427FR
This item is not returnable.
View return policy
Description
Nanog Monoclonal antibody specifically detects Nanog in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Nanog | |
| Monoclonal | |
| DyLight 680 | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| Purified recombinant fragment of human Nanog (amino acids 20-166) expressed in E. coli. [UniProt# Q9H9S0] | |
| 0.1 mL | |
| Cellular Markers, Stem Cell Markers | |
| 79923 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 1E6C4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction