missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanog Antibody (5A10), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04320UV
This item is not returnable.
View return policy
Description
Nanog Monoclonal specifically detects Nanog in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Nanog | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NANOG | |
| Recombinant human Nanog (1-154aa) purified from E. coli (NP_079141). | |
| 0.1 mL | |
| Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers | |
| 79923 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 5A10 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human Nanog mAb, clone 5A10, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Nanog protein. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction