missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanog Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-588R
This item is not returnable.
View return policy
Description
Nanog Polyclonal specifically detects Nanog in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Nanog | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| FLJ12581, FLJ40451, hNanog, homeobox protein NANOG, Homeobox transcription factor Nanog, homeobox transcription factor Nanog-delta 48, Nanog homeobox | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB100-588 is specific for the Nanog protein. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NANOG | |
| A synthetic peptide which maps to a region between residues 250 and the C-terminus (residue 305) of mouse Nanog homeobox using the numbering given in TrEMBL entry Q80Z64 (GeneID 71950). | |
| 100 μL | |
| Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers | |
| 79923 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction