Learn More
NANOS1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98705MFV610
Product Code. 18564271
  • € 451.00 / 0.10mL
Description
NANOS1 Polyclonal antibody specifically detects NANOS1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NANOS1
mFluor Violet 610 SE
Store at 4C in the dark.
Polyclonal
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin