missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanos2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76370PCP
This item is not returnable.
View return policy
Description
Nanos2 Polyclonal antibody specifically detects Nanos2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| Nanos2 | |
| Polyclonal | |
| Western Blot, ELISA | |
| nanos homolog 2, nanos homolog 2 (Drosophila), NOS2NOS-2 | |
| Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of the human Nanos2. The immunogen is located within the last 50 amino acids of Nanos2. Amino Acid Squence: QSLYRRSGRNSAGRR | |
| 0.1 mL | |
| Signal Transduction | |
| 339345 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction