missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NANP Antibody (OTI4D11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72870AF405
This item is not returnable.
View return policy
Description
NANP Monoclonal specifically detects NANP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| NANP | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NANP | |
| Full length recombinant protein of human NANP (NP_689880) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI4D11 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| C20orf147, chromosome 20 open reading frame 147, dJ694B14.3, EC 3.1.3.29, haloacid dehalogenase-like hydrolase domain containing 4, Haloacid dehalogenase-like hydrolase domain-containing protein 4, HDHD4Neu5Ac-9-Pase, MGC26833, N-acetylneuraminic acid phosphatase, N-acylneuraminate-9-phosphatase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 140838.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction