missing translation for 'onlineSavingsMsg'
Learn More

NANP Monoclonal Antibody (OTI4F5), TrueMAB™, OriGene

Product Code. 17552002
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17552002 30 μL 30µL
17532002 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17552002 Supplier Origene Technologies Supplier No. TA504598S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NANP Monoclonal antibody specifically detects NANP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NANP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NANP
Gene Accession No. Q8TBE9
Gene Alias 1600031M04Rik; C20orf147; dJ694B14.3; haloacid dehalogenase-like hydrolase domain containing 4; haloacid dehalogenase-like hydrolase domain-containing protein 4; Hdhd4; hypothetical protein LOC553795; N-acetylneuraminic acid phosphatase; N-acylneuraminate-9-phosphatase; NANP; nanp {ECO:0000250; Neu5Ac-9-Pase; RGD1306009; UniProtKB:Q9CPT3}; zgc:111947
Host Species Mouse
Immunogen Full length human recombinant protein of human NANP produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140838
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.