missing translation for 'onlineSavingsMsg'
Learn More

NAP1L4 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-36542CL1

Product Code. 30147733

  • € 1020.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NAP1L4 Polyclonal antibody specifically detects NAP1L4 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NAP1L4
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Immunoprecipitation, Western Blot (Negative)
CoraFluor 1
hNAP2, MGC4565, NAP-2, NAP2L, NAP2NAP1L4b, nucleosome assembly protein 1-like 4, nucleosome assembly protein 1-like 4b, Nucleosome assembly protein 2
The immunogen recognized by the antibody maps to a region between residue 125 to 175 of human Nucleosome Assembly Protein 2 using the numbering given in entry NP_005960.1.
0.1 mL
Chromatin Modifiers
4676
Store at 4°C in the dark. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.