missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAP1L5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94432-0.1ml
This item is not returnable.
View return policy
Description
NAP1L5 Polyclonal antibody specifically detects NAP1L5 in Human, Mouse samples. It is validated for Western Blot
Specifications
| NAP1L5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| down-regulated in liver malignancy, DRLM1110020M21Rik, nucleosome assembly protein 1-like 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-120 of human NAP1L5 (NP_715638.1). GGAQGGDCDSAAGDPDSAAGQMAEEPQTPAENAPKPKNDFIESLPNSVKCRVLALKKLQKRCDKIEAKFDKEFQALEKKYNDIYKPLLAKI | |
| 0.1 mL | |
| Cell Biology | |
| 266812 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction