missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPE-PLD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48571
This item is not returnable.
View return policy
Description
NAPE-PLD Polyclonal antibody specifically detects NAPE-PLD in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NAPE-PLD | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| C7orf18, DKFZp781D1098, EC 3.1.4, EC 3.1.4.-, N-acyl phosphatidylethanolamine phospholipase DNAPE-PLDFMP30, N-acyl-phosphatidylethanolamine-hydrolyzing phospholipase D, NAPE-hydrolyzing phospholipase D | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ELPVLKPYFITNPEEAGVREAGLRVTWLGHATVMVEMDELIFLTDPIFSSRASPSQYMGPKRFRRSPCTISELPPIDAVLISHNHYDHLDYNSV | |
| 0.1 mL | |
| Lipid and Metabolism, Signal Transduction | |
| 222236 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction