missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1-carbonitrile, 95%

Product Code. p-7046475
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11333556

Brand: Thermo Scientific Alfa Aesar L14498.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-53-3 | C11H7N | 153.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-53-3
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.184
MDL Number MFCD00003866
InChI Key YJMNOKOLADGBKA-UHFFFAOYSA-N
Synonym 1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd
PubChem CID 6846
IUPAC Name naphthalene-1-carbonitrile
SMILES C1=CC=C2C(=C1)C=CC=C2C#N

Specifications

Melting Point 33°C to 38°C
Boiling Point 299°C
Quantity 5 g
UN Number UN3439
Beilstein 971255
Formula Weight 153.18
Percent Purity 95%
Chemical Name or Material Naphthalene-1-carbonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.