missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthalene-2,3-dicarboxaldehyde, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Naphthalene-2,3-Dicarboxaldehyde |
| CAS | 7149-49-7 |
| Molecular Formula | C12 H8 O2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C12H8O2/c13-7-11-5-9-3-1-2-4-10(9)6-12(11)8-14/h1-8H |
| SMILES | O=Cc1cc2ccccc2cc1C=O |
| IUPAC Name | naphthalene-2,3-dicarbaldehyde |
| Molecular Weight (g/mol) | 184.19 |
| Formula Weight | 184.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction