Learn More
NaPi-IIa Rabbit, anti-Human, Polyclonal, Proteintech
Product Code. 18076395 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18076395 150 μL 150µL
18064795 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18076395 Supplier Proteintech Supplier No. 267831AP150UL
Rabbit Polyclonal Antibody
The NaPi-IIa antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NaPi-IIa. This antibody recognizes human, mouse, rat antigen. The NaPi-IIa antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen NaPi-IIa
Applications Western Blot, ELISA
Concentration 750 μg/mL
Gene Symbols Na(+)/Pi cotransporter 2A, NaPi 2a, NAPI 3, napi2a, NaPi-IIa, SLC11, SLC17A2, SLC34A1
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 6569
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG