missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPSA Monoclonal Antibody (OTI3E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808666S
This item is not returnable.
View return policy
Description
NAPSA Monoclonal antibody specifically detects NAPSA in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)Specifications
| NAPSA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10909 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1191 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| OTI3E5 | |
| Unconjugated | |
| Clu | |
| asp 4; ASP4; Aspartyl protease 4; KAP; Kdap; KDAP-1; kidney-derived aspartic protease-like protein; Nap; NAP1; NAPA; Napsa; napsin A aspartic peptidase; napsin-1; Napsin-A; pronapsin; pronapsin A; SNAPA; TA01/TA02 | |
| Human recombinant protein fragment corresponding to amino acids 23-227 of human CLU produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction