missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPSA Monoclonal Antibody (OTI3E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808666
This item is not returnable.
View return policy
Description
Napsin 1 is an aspartic protease that is generally expressed in the lung, but also present in kidney and in metastatic carcinomas throughout the body. Napsin 1 processes surfactant protein B in healthy pulmonary and renal tissues, but is abnormally regulated when carcinogenesis occurs. Thus, many researchers have found napsin evaluation to be useful in identifying several cancers, and differentiating primary tumors from metastases. When combined with thyroid transcription factor analysis, napsin can be used as a specific biomarker for differentiation of carcinoma types. Napsin has also been associated with pulmonary alveolar proteinosis as well as in protein catabolism in renal tubules.Specifications
| NAPSA | |
| Monoclonal | |
| Unconjugated | |
| NAPSA | |
| KAP, Kdap, NAP1, NAPA, SNAPA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9476 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| OTI3E5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O96009 | |
| NAPSA | |
| Human recombit protein fragment corresponding to amino acids 64-244 of human NAPSA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction