missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Napsin A Antibody (NAPSA/7165R), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20977B
This item is not returnable.
View return policy
Description
Napsin A Monoclonal antibody specifically detects Napsin A in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Napsin A | |
| Monoclonal | |
| Biotin | |
| Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02 | |
| Recombinant fragment (aa50-150) of human Napsin A protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers | |
| 9476 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| NAPSA/7165R | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction