missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Napsin A Rabbit anti-Human, Clone: NAPSA/4400R, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 528.00
Specifications
| Antigen | Napsin A |
|---|---|
| Clone | NAPSA/4400R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Napsin A Monoclonal specifically detects Napsin A in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| Napsin A | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 9476 | |
| IgG | |
| Protein A purified |
| NAPSA/4400R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02 | |
| Recombinant fragment (aa50-150) of human Napsin A protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title