missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Narciclasine

Click to view available options
:
1mg
Description
Narciclasine is an exhibits antiproliferative and pro-apoptotic effects in carcinoma cells and displays cytotoxic activity against a panel of 60 cancer cell lines (mean IC50 = 47 nM). Activity decreases rate of cell division and increases mitosis duration in vitro. Also modulates the Rho/ROCK/LIM kinase/cofilin pathway; stimulates RhoA activation and induces actin polymerization.
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | Other Rho GTPase Compounds |
| Purity | >98% |
| Molecular Weight (g/mol) | 307.26 |
| Inhibitors | Narciclasine |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction