missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARG1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23845-1-AP-150UL
This item is not returnable.
View return policy
Description
N-alpha-acetyltransferase 15, NatA auxiliary subunit also known as gastric cancer antigen Ga19 (GA19), NMDA receptor-regulated protein 1 (NARG1), and Tbdn100 is a protein that in humans is encoded by the NAA15 gene.NARG1 is the auxiliary subunit of the NatA (Nalpha-acetyltransferase A) complex. This NatA complex can associate with the ribosome and catalyzes the transfer of an acetyl group to the Nalpha-terminal amino group of proteins as they emerge from the exit tunnel.Specifications
| NARG1 | |
| Polyclonal | |
| Unconjugated | |
| NAA15 | |
| Ga19, Gastric cancer antigen Ga19, N terminal acetyltransferase, NAA15, NARG1, NATH, Protein tubedown 1, TBDN100 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80155 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BXJ9 | |
| NAA15 | |
| NARG1 Fusion Protein Ag20842 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction