Learn More
Schneider Gerd™ Narrow Double Spatula
Product Code. 10021714
Click to view available options
Length (Metric):
350 mm
400 mm
Unit Size:
Each
Product Code. 10021714
Brand: Schneider Gerd™ 308
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Shape Narrow
Product Type Double Ended Spatula
Width (Metric) Blade 16 mm
Dimensions (L x W) 400 x 16 mm
Length (Metric) 400 mm