Learn More
BRAND™ Narrow Neck Erlenmeyer Flask
Narrow Neck Erlenmeyer Flask features a beaded rim and graduation marks.
Brand: BRAND™ 92714
102.54 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.29000kg
Description
- Borosilicate Glass 3.3
- DIN ISO 1773
- With beaded rim and graduation
Specifications
25 mL | |
Narrow | |
Beaded |
Borosilicate Glass (3.3) | |
Yes |