Learn More
Brand™ Narrow Neck LDPE Bottles with Cap
Manufactured from flexible, translucent, LDPE. BRAND™ Narrow Neck LDPE Bottles with Cap are economical, and can serve many purposes. Available in a variety of capacities.
€ 4.25 - € 124.00
Specifications
Closure Type | Screw Cap |
---|---|
Product Type | Bottle |
For Use With (Equipment) | Multi-purpose bottle |
Material | LDPE |
Transparency | Translucent |
Product Code | Brand | Capacity (Metric) | Thread Size (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Thread Size (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||
15221299
|
Brand™
129428 |
50 mL | GL 18 | 39 mm | 85 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15241299
|
Brand™
129448 |
250 mL | GL 25 | 59 mm | 140 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15251299
|
Brand™
129454 |
500 mL | GL 25 | 75 mm | 180 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10422291
|
Brand™
129462 |
1000 mL | GL 28 | 94 mm | 212 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10471914
|
Brand™
129462 |
1000 mL | GL 28 | 94 mm | 212 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15231299
|
Brand™
129438 |
100 mL | GL 18 | 45 mm | 106 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Manufactured from flexible, translucent, LDPE.
- Economical, multi-purpose bottle includes cap.
- Available in a variety of capacities.
Specifications
Screw Cap | |
Multi-purpose bottle | |
Translucent | |
Narrow |
Bottle | |
LDPE | |
Yes |