missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duran™ Narrow Neck Soda Lime Glass Reagent Bottles
Click to view available options
Capacity (Metric):
100 mL
Specifications
Diameter (Metric) | 52 mm |
Height (Metric) | 96 mm |
Product Type | Reagent Bottle |
Capacity (Metric) | 100 mL |
Color | Clear |
For Use With (Application) | Suitable for storage of aggressive media, which could attack plastic parts. |
Includes | Standard Ground Joint |
Material | Soda Lime Glass |
Neck ID (Metric) | 14/15 |
Quantity | 10Pack |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'