missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARS Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98529DL594
This item is not returnable.
View return policy
Description
NARS Polyclonal antibody specifically detects NARS in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NARS | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| ASNRS, asparagine tRNA ligase 1, cytoplasmic, Asparagine--tRNA ligase, asparaginyl-tRNA synthetase, asparaginyl-tRNA synthetase, cytoplasmic, EC 6.1.1.22, NARS1 | |
| Produced in rabbits immunized with E. coli-derived Human NARS fragment. | |
| 0.1 mL | |
| Epigenetics | |
| 4677 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction