missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NARS Polyclonal Antibody

Description
Positive Control: HepG2 cell lysate, K562 cell lysate, 293 cell lysate, NIH/3T3 cell lysate, mouse liver tissue lysate, mouse brain tissue, mouse cerebellum tissue, rat cerebellum tissue. Subcellular Location: Cytoplasm.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Asparaginyl-tRNA synthetase is localized to the cytoplasm and belongs to the class II family of tRNA synthetases. The N-terminal domain represents the signature sequence for the eukaryotic asparaginyl-tRNA synthetases.
Specifications
Specifications
| Antigen | NARS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | NARS1 |
| Gene Accession No. | O43776, Q8BP47 |
| Gene Alias | 3010001M15Rik; AA960128; AsnRS; asparagine tRNA ligase 1, cytoplasmic; asparagine--tRNA ligase, cytoplasmic; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase, cytoplasmic; C78150; LRRGT00113; Nars; NARS1 |
| Gene Symbols | NARS1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?