missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARS2 Antibody (OTI10C10), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72876PE
This item is not returnable.
View return policy
Description
NARS2 Monoclonal antibody specifically detects NARS2 in Human samples. It is validated for Western Blot
Specifications
| NARS2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 79731 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI10C10 | |
| Western Blot | |
| asnRS, asparagine tRNA ligase 2, mitochondrial (putative), Asparagine--tRNA ligase, asparaginyl-tRNA synthetase 2, mitochondrial (putative), EC 6.1.1, EC 6.1.1.22, FLJ23441, probable asparaginyl-tRNA synthetase, mitochondrial, SLM5 | |
| Full length human recombinant protein of human NARS2 (NP_078954) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction