Learn More
NARS2 Monoclonal Antibody (OTI10E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA806499S
Encompass_Preferred
 View more versions of this product
Product Code. 17526673
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
NARS2 Monoclonal antibody specifically detects NARS2 in Human samples. It is validated for Western Blot
Specifications
Specifications
NARS2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96I59
Mouse
Affinity chromatography
RUO
79731
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI10E2
Unconjugated
NARS2
AI875199; asnRS; asparagine tRNA ligase 2, mitochondrial (putative); asparagine--tRNA ligase; Asparaginyl-tRNA synthetase; asparaginyl-tRNA synthetase 2 (mitochondrial)(putative); asparaginyl-tRNA synthetase 2, mitochondrial; asparaginyl-tRNA synthetase 2, mitochondrial (putative); deafness, autosomal recessive 94; DFNB94; EC 6.1.1.22; Nars2; Nars2, similar to CG6796-PA; probable asparagine--tRNA ligase, mitochondrial; probable asparaginyl-tRNA synthetase, mitochondrial; RGD1305145; SLM5
Full length human recombinant protein of human NARS2 produced in HEK293T cell.
30 μL