Learn More
NARS2 Mouse anti-Human, Clone: OTI10E2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806499
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Asparagine-tRNA ligase (EC 6.1.1.22) catalyzes the chemical reaction: ATP + L-asparagine + tRNAAsn <--> AMP + diphosphate + L-asparaginyl-tRNAAsn. This enzyme participates in alanine and aspartate metabolism and aminoacyl-tRNA biosynthesis.Specifications
| NARS2 | |
| Monoclonal | |
| Unconjugated | |
| NARS2 | |
| DFNB94, SLM5, asnRS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79731 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI10E2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96I59 | |
| NARS2 | |
| Full length human recombit protein of human NARS2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.