missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAT10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15033
This item is not returnable.
View return policy
Description
NAT10 Polyclonal specifically detects NAT10 in Human samples. It is validated for Western Blot.
Specifications
| NAT10 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
| NAT10 | |
| Recombinant protein encompassing a sequence within the center region of human NAT10. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 55226 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| ALP, DKFZp434C116, EC 2.3.1.-, FLJ10774, FLJ12179, FLJ23850, hALP, KIAA1709, N-acetyltransferase 10, N-acetyltransferase 10 (GCN5-related), N-acetyltransferase-like protein, NET43 | |
| Rabbit | |
| 116 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (98%), Bovine (98%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction