missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAT10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17171194 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17171194 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17171194 Supplier Invitrogen™ Supplier No. PA5113336

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NAT10 Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA

IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT10
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 3.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Nat10
Gene Accession No. Q9H0A0
Gene Alias 18S rRNA cytosine acetyltransferase; AI429152; ALP; hALP; KIAA1709; N-acetyltransferase 10; N-acetyltransferase 10 (GCN5-related); N-acetyltransferase-like protein; NAT10; NET43; novel putative ATPase domain containing protein; RGD1306717; RNA cytidine acetyltransferase
Gene Symbols Nat10
Host Species Rabbit
Immunogen Recombinant Human RNA cytidine acetyltransferase protein (909-1025AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55226
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.