missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAT10 Polyclonal Antibody

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5142262

Product Code. 17838282

  • € 474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NAT10
Polyclonal
Unconjugated
Nat10
18S rRNA cytosine acetyltransferase; AI429152; ALP; hALP; KIAA1709; N-acetyltransferase 10; N-acetyltransferase 10 (GCN5-related); N-acetyltransferase-like protein; NAT10; NET43; novel putative ATPase domain containing protein; RGD1306717; RNA cytidine acetyltransferase
Goat
Ammonium sulfate precipitation
RUO
55226
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q9H0A0
Nat10
Peptide with sequence C-KSDKKRKLEAKQ.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.