Learn More
NAT10 Rabbit, anti-Human, CoraLite Plus 488, Clone: 5B9, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech CL488-82585-100UL
 View more versions of this product
Product Code. 18007815
  • € 450.00 / 100µL
Description
The NAT10 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human NAT10. This antibody recognizes human antigen. The NAT10 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
NAT10
5B9
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa
IgG
Immunofluorescence
1000 μg/mL
ALP, DKFZp434C116, FLJ10774, FLJ12179, FLJ23850, KIAA1709, N acetyltransferase 10, NAT10
100 μL
55226
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.