missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAT10 Recombinant Rabbit Monoclonal Antibody (K01_1K48)

Product Code. 30828185
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828185 50 μL
30829184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828185 Supplier Invitrogen™ Supplier No. MA564653

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NAT10 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT10
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1K48
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene Nat10
Gene Accession No. Q9H0A0
Gene Alias 18S rRNA cytosine acetyltransferase; AI429152; ALP; hALP; KIAA1709; N-acetyltransferase 10; N-acetyltransferase 10 (GCN5-related); N-acetyltransferase-like protein; NAT10; NET43; novel putative ATPase domain containing protein; RGD1306717; RNA cytidine acetyltransferase
Gene Symbols Nat10
Host Species Rabbit
Immunogen Recombinant protein of human NAT10.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311257, 55226
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.