missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Semaphorin 6A Monoclonal antibody specifically detects Semaphorin 6A in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Semaphorin 6A |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 001 |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | KIAA1368HT018, SEMA, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Sema VIA, SEMA6A1, SEMA6A-1, semaphorin 6A-1, Semaphorin VIA, semaphorin-6A, Semaphorin-6A-1, SEMAQ, VIA |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Semaphorin 6A (NP_065847.1; Met1-Thr649). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?