Learn More
Invitrogen™ NAT14 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5115617
Product Code. 17298393
  • € 498.00 / 100µL
Description
Antibody detects endogenous levels of total NAT14.
Probable acetyltransferase that binds the 5'-GGACTACAG-3' sequence of coproporphyrinogen oxidase promoter. Able to activate transcription of a reporter construct in vitro.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NAT14
Polyclonal
Unconjugated
NAT14
BC047219; K562 cell-derived leucine-zipper-like protein 1; K562 cells-derived leucine zipper-like protein 1; K562 cells-derived leucine-zipper-like protein 1; KLP1; N-acetyltransferase 14; N-acetyltransferase 14 (GCN5-related, putative); N-acetyltransferase 14 (putative); Nat14; RGD1565991; Zfp628; zinc finger protein 628
Rabbit
Affinity chromatography
RUO
269854, 361500, 57106
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
Q8BVG8, Q8WUY8
NAT14
A synthesized peptide derived from human NAT14(Accession Q8WUY8), corresponding to amino acid residues A163-L206.
100 μL
Primary
Human, Mouse, Rat