Learn More
Invitrogen™ NAT14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115617
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NAT14.
Probable acetyltransferase that binds the 5'-GGACTACAG-3' sequence of coproporphyrinogen oxidase promoter. Able to activate transcription of a reporter construct in vitro.
Specifications
| NAT14 | |
| Polyclonal | |
| Unconjugated | |
| NAT14 | |
| BC047219; K562 cell-derived leucine-zipper-like protein 1; K562 cells-derived leucine zipper-like protein 1; K562 cells-derived leucine-zipper-like protein 1; KLP1; N-acetyltransferase 14; N-acetyltransferase 14 (GCN5-related, putative); N-acetyltransferase 14 (putative); Nat14; RGD1565991; Zfp628; zinc finger protein 628 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 269854, 361500, 57106 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8BVG8, Q8WUY8 | |
| NAT14 | |
| A synthesized peptide derived from human NAT14(Accession Q8WUY8), corresponding to amino acid residues A163-L206. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |