missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAT15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559351
This item is not returnable.
View return policy
Description
Immunogen sequence: EVVPSSALSE VSLRLLCHDD IDTVKHLCGD WFPIEYPDSW YRDITSNKKF FSLAATYRGA IVGMIVAEIK NRTKI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Histone acetyltransferase localized in the Golgi apparatus that mediates acetylation of free histone H4, thereby facilitating nucleosome assembly. Has a preference for free histone H4 'Lys-20'(H4K20ac), 'Lys-79'(H4K79ac) and 'Lys-91' (H4K91ac). Also displays alpha (N-terminal) acetyltransferase activity towards a range of N-terminal sequences including those starting with Met-Lys, Met-Val, Met-Ala and Met-Met. Required for normal chromosomal segregation during anaphase.
Specifications
| NAT15 | |
| Polyclonal | |
| Unconjugated | |
| NAA60 | |
| 1200013P24Rik; AI315146; GNAT acetyltransferase; GNAT acetytransferase; HAT4; Histone acetyltransferase type B protein 4; hNaa60; N(alpha)-acetyltransferase 60, NatF catalytic subunit; Naa60; N-acetyltransferase 15; N-acetyltransferase 15 (GCN5-related, putative); N-alpha-acetyltransferase 60; N-alpha-acetyltransferase 60, NatF catalytic subunit; N-alpha-acetyltransferase F; NAT15; NatF; natF catalytic subunit; RGD1308915; UNQ2771/PRO7155 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79903 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7X0 | |
| NAA60 | |
| Recombinant protein corresponding to Human NAT15. Recombinant protein control fragment (Product #RP-98652). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction