Learn More
Invitrogen™ NAT8 Monoclonal Antibody (OTI6H3)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA524948
Product Code. 15944554
  • € 572.00 / 100µL
Description
NAT8 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This protein, isolated using the differential display method to detect tissue-specific genes, is specifically expressed in kidney and liver. The encoded protein shows amino acid sequence similarity to N-acetyltransferases. A similar protein in Xenopus affects cell adhesion and gastrulation movements, and may be localized in the secretory pathway. A highly similar paralog is found in a cluster with this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NAT8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHE5
NAT8
Full-length protein expressed in 293T cell transfected with human NAT8 expression vector.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI6H3
Unconjugated
NAT8
0610037O16Rik; Acetyltransferase 2; ATase2; camello-like 4; camello-like protein 1; Camello-like protein 4; CCNAT; CML1; Cml4; Cysteinyl-conjugate N-acetyltransferase; GLA; Hcml1; kidney- and liver-specific gene product; N-acetyltransferase 8; N-acetyltransferase 8 (camello like); N-acetyltransferase 8 (GCN5-related, putative); N-acetyltransferase 8 (putative); NAT8; probable N-acetyltransferase 8; probable N-acetyltransferase CML4; putative N-acetyltransferase Camello 4; TSC501; TSC510
Mouse
Affinity Chromatography
RUO
9027
-20°C, Avoid Freeze/Thaw Cycles