missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Natalizumab Chimeric Recombinant Rabbit Monoclonal Antibody (Natalizumab)
GREENER_CHOICE

Product Code. 17911281
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911281 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17911281 Supplier Invitrogen™ Supplier No. MA548046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody binds to the alpha 4 binds subunit of alpha-4Beta1 (also known as very late antigen 4 [VLA-4] or CD49d-CD29) and alpha-4Beta7 integrin.

Natalizumab is a recombinant, humanized monoclonal antibody, binds to alpha4beta1-integrinand blocks its interaction with vascular cell adhesion molecule-1 (VCAM-1). Natalizumab can be used for the treatment of relapsing remitting multiple sclerosis and Crohn's disease. Natalizumab is also the first targeted therapy which blocks an essential mechanism for lymphocyte entry to the CNS and thus prevents acute demyelinating relapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Natalizumab Chimeric
Applications Flow Cytometry, Inhibition Assays
Classification Recombinant Monoclonal
Clone Natalizumab
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias AN 100226; BG00002
Host Species Rabbit
Immunogen The RAMOS cell line, a non-EBV releasing cell line, injected into mice.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.